Learn More
Biomatik Corporation DA (Dopamine) ELISA Kit, 96 tests
Catalog No. 501501130
Encompass_Preferred
The DA (Dopamine) ELISA Kit allows for quantitative determination of DA in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1130 Supplier Biomatik Corporation Supplier No. EKF57981
The DA (Dopamine) ELISA Kit allows for quantitative determination of DA in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.