Learn More
Biomatik Corporation T (Testosterone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57983
Encompass_Preferred
T (Testosterone) ELISA Kit for detecting T in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Testosterone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1132