Learn More
Biomatik Corporation T3 (Triiodothyronine) ELISA Kit, 96 tests
Catalog No. 501501134
Encompass_Preferred
T3 (Triiodothyronine) ELISA Kit for detecting T3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: T3; Triiodothyronine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1134 Supplier Biomatik Corporation Supplier No. EKF57985
T3 (Triiodothyronine) ELISA Kit for detecting T3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: T3; Triiodothyronine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.