Learn More
Biomatik Corporation VB12 (Vitamin B12) ELISA Kit, 96 tests
Catalog No. 501501135
Encompass_Preferred
VB12 (Vitamin B12) ELISA Kit for detecting VB12 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 50-500pg/ml. Sensitivity: 25pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Vitamin B12; VB12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1135 Supplier Biomatik Corporation Supplier No. EKF57986
VB12 (Vitamin B12) ELISA Kit for detecting VB12 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 50-500pg/ml. Sensitivity: 25pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Vitamin B12; VB12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.