Learn More
Biomatik Corporation OT (Oxytocin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57988
Encompass_Preferred
OT (Oxytocin) ELISA Kit for detecting OT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: OT; Oxytocin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1137