Learn More
Biomatik Corporation DHT (Dihydrotestosterone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57989
Encompass_Preferred
The DHT (Dihydrotestosterone) ELISA Kit allows for quantitative determination of DHT in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 39.063-2500 pg/mL. Sensitivity: 23.438 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1138