Learn More
Biomatik Corporation HA (Hyaluronic Acid) ELISA Kit, 96 tests
Catalog No. 501501139
Encompass_Preferred
The HA (Hyaluronic Acid) ELISA Kit allows for quantitative determination of HA in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 1.563-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1139 Supplier Biomatik Corporation Supplier No. EKF57990
The HA (Hyaluronic Acid) ELISA Kit allows for quantitative determination of HA in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 1.563-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.