Learn More
Biomatik Corporation 3-NT (3-Nitrotyrosine) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57991
Encompass_Preferred
The 3-NT (3-Nitrotyrosine) ELISA Kit allows for quantitative determination of 3-NT in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 1.563-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1140