Learn More
Biomatik Corporation MDA (Malondialdehyde) ELISA Kit, 96 tests
Catalog No. 501501145
Encompass_Preferred
MDA (Malondialdehyde) ELISA Kit for detecting MDA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 7.813-500ng/ml. Sensitivity: 4.688ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: MDA; Malondialdehyde. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1145 Supplier Biomatik Corporation Supplier No. EKF57996
MDA (Malondialdehyde) ELISA Kit for detecting MDA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 7.813-500ng/ml. Sensitivity: 4.688ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: MDA; Malondialdehyde. Restrictions: For Research Use Only. Not for use in diagnostic procedures.