Learn More
Biomatik Corporation CORT (Corticosterone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57998
Encompass_Preferred
The CORT (Corticosterone) ELISA Kit allows for quantitative determination of CORT in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 2.81-180 ng/mL. Sensitivity: 1.688 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1147