Learn More
Biomatik Corporation Rat Adrenocorticotropic Hormone (ACTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54008
Encompass_Preferred
Rat Adrenocorticotropic Hormone (ACTH) ELISA Kit for detecting ACTH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 4.76pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 4h, 30min. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1149