Learn More
Biomatik Corporation Human Angiotensin II Receptor 1 (AGTR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54031
Encompass_Preferred
The Human Angiotensin II Receptor 1 (AGTR1) ELISA Kit allows for quantitative determination in tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P30556. NCBI Gene ID: 185. Restriction: For Research Use Only.
Catalog No. 50-150-1166