Learn More
Biomatik Corporation Rat Anti-Mullerian Hormone (AMH) ELISA Kit, 96 tests
Catalog No. 501501173
Encompass_Preferred
Rat Anti-Mullerian Hormone (AMH) ELISA Kit for detecting AMH in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P49000. Gene ID: 25378. Target Synonyms: MIF; MIH; MIS; Mullerian Inhibiting Factor; Mullerian Inhibiting Hormone; Mullerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1173 Supplier Biomatik Corporation Supplier No. EKL54044
Rat Anti-Mullerian Hormone (AMH) ELISA Kit for detecting AMH in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P49000. Gene ID: 25378. Target Synonyms: MIF; MIH; MIS; Mullerian Inhibiting Factor; Mullerian Inhibiting Hormone; Mullerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.