Learn More
Biomatik Corporation Rat Angiotensin I (AngI) ELISA Kit, 96 tests
Catalog No. 501501178
Encompass_Preferred
Rat Angiotensin I (AngI) ELISA Kit for detecting AngI in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 4.62pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Target Synonyms: Ang-I; Angiotensin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1178 Supplier Biomatik Corporation Supplier No. EKL54054
Rat Angiotensin I (AngI) ELISA Kit for detecting AngI in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 4.62pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Target Synonyms: Ang-I; Angiotensin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.