Learn More
Biomatik Corporation Human B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit, 96 tests
Catalog No. 501501186
Encompass_Preferred
The Human B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.57 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P10415. NCBI Gene ID: 596. Restriction: For Research Use Only.
Catalog No. 50-150-1186 Supplier Biomatik Corporation Supplier No. EKL54090
The Human B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.57 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P10415. NCBI Gene ID: 596. Restriction: For Research Use Only.