Learn More
Biomatik Corporation Rat Brain Derived Neurotrophic Factor (BDNF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54103
Encompass_Preferred
Rat Brain Derived Neurotrophic Factor (BDNF) ELISA Kit for detecting BDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P23363. Gene ID: 24225. Target Synonyms: Neurotrophin; Abrineurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1192