Learn More
Biomatik Corporation Bovine Bone Morphogenetic Protein 2 (BMP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54111
Encompass_Preferred
The Bovine Bone Morphogenetic Protein 2 (BMP2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine Cattle. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.107 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: A5PJI9. NCBI Gene ID: 615037. Restriction: For Research Use Only.
Catalog No. 50-150-1195