Learn More
Biomatik Corporation Rat Bone Morphogenetic Protein 2 (BMP2) ELISA Kit, 96 tests
Catalog No. 501501199
Encompass_Preferred
Rat Bone Morphogenetic Protein 2 (BMP2) ELISA Kit for detecting BMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 25.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P49001. Gene ID: 29373. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1199 Supplier Biomatik Corporation Supplier No. EKL54116
Rat Bone Morphogenetic Protein 2 (BMP2) ELISA Kit for detecting BMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 25.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P49001. Gene ID: 29373. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.