Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 4 (BMP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54123
Encompass_Preferred
Mouse Bone Morphogenetic Protein 4 (BMP4) ELISA Kit for detecting BMP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P21275. Gene ID: 12159. Target Synonyms: BMP2B; BMP2-B; BMP2B1; ZYME; DVR4; Bone morphogenetic protein 2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1202