Learn More
Biomatik Corporation Rat Bone Morphogenetic Protein 4 (BMP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54125
Encompass_Preferred
Rat Bone Morphogenetic Protein 4 (BMP4) ELISA Kit for detecting BMP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q06826. Gene ID: 25296. Target Synonyms: BMP2B; BMP2-B; BMP2B1; ZYME; DVR4; Bone morphogenetic protein 2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1203