Learn More
Biomatik Corporation Human Bone Morphogenetic Protein Receptor 1A (BMPR1A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54127
Encompass_Preferred
The Human Bone Morphogenetic Protein Receptor 1A (BMPR1A) ELISA Kit allows for quantitative determination in tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.11 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P36894. NCBI Gene ID: 657. Restriction: For Research Use Only.
Catalog No. 50-150-1204