Learn More
Biomatik Corporation Rat Caspase 3 (CASP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54148
Encompass_Preferred
Rat Caspase 3 (CASP3) ELISA Kit for detecting CASP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.49ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P55213. Gene ID: 25402. Target Synonyms: CPP32; CPP32B; SCA1; Apoptain; Yama; Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 3; SREBP cleavage activity 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1211