Learn More
Biomatik Corporation Human Cholecystokinin (CCK) ELISA Kit, 96 tests
Catalog No. 501501212
Encompass_Preferred
The Human Cholecystokinin (CCK) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 4.55 pg/mL. Format: 96-Well Plate. Assay type: competitive inhibition. UniProt ID: P06307. NCBI Gene ID: 885. Restriction: For Research Use Only.
Catalog No. 50-150-1212 Supplier Biomatik Corporation Supplier No. EKL54157
The Human Cholecystokinin (CCK) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 4.55 pg/mL. Format: 96-Well Plate. Assay type: competitive inhibition. UniProt ID: P06307. NCBI Gene ID: 885. Restriction: For Research Use Only.