Learn More
Biomatik Corporation Rat Carboxypeptidase B1, Tissue (CPB1) ELISA Kit, 96 tests
Catalog No. 501501213
Encompass_Preferred
Rat Carboxypeptidase B1, Tissue (CPB1) ELISA Kit for detecting CPB1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P19223. Gene ID: 24271. Target Synonyms: CPB; PCPB; PASP; Pancreas-specific protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1213 Supplier Biomatik Corporation Supplier No. EKL54175
Rat Carboxypeptidase B1, Tissue (CPB1) ELISA Kit for detecting CPB1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P19223. Gene ID: 24271. Target Synonyms: CPB; PCPB; PASP; Pancreas-specific protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.