Learn More
Biomatik Corporation Rat Connective Tissue Growth Factor (CTGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54190
Encompass_Preferred
Rat Connective Tissue Growth Factor (CTGF) ELISA Kit for detecting CTGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400pg/ml. Sensitivity: 2.46pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q9R1E9. Gene ID: 64032. Target Synonyms: CCN2; HCS24; IGFBP8; NOV2; Insulin-Like Growth Factor-Binding Protein 8; Hypertrophic Chondrocyte-Specific Protein 24; CCN Family Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1217