Learn More
Biomatik Corporation Human Cytotoxic T-Lymphocyte Associated Antigen 4 (CTLA4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54192
Encompass_Preferred
The Human Cytotoxic T-Lymphocyte Associated Antigen 4 (CTLA4) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P16410. NCBI Gene ID: 1493. Restriction: For Research Use Only.
Catalog No. 50-150-1218