Learn More
Biomatik Corporation Human Cyclophilin A (CYPA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54198
Encompass_Preferred
The Human Cyclophilin A (CYPA) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.124 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P62937. NCBI Gene ID: 5478. Restriction: For Research Use Only.
Catalog No. 50-150-1219