Learn More
Biomatik Corporation Mouse Endothelin 1 (EDN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54214
Encompass_Preferred
Mouse Endothelin 1 (EDN1) ELISA Kit for detecting EDN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.812-500pg/ml. Sensitivity: 2.45pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 4h, 30min. Accession Number: P22387. Gene ID: 13614. Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1221