Learn More
Biomatik Corporation Human Epidermal Growth Factor Receptor 2 (EGFR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54231
Encompass_Preferred
The Human Epidermal Growth Factor Receptor 2 (EGFR2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.781-50 ng/mL. Sensitivity: 0.25 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P04626. NCBI Gene ID: 2064. Restriction: For Research Use Only.
Catalog No. 50-150-1232