Learn More
Biomatik Corporation Human Elastin (ELN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54236
Encompass_Preferred
The Human Elastin (ELN) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.468-30 ng/mL. Sensitivity: 0.21 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P15502. NCBI Gene ID: 2006. Restriction: For Research Use Only.
Catalog No. 50-150-1234