Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 10 (FGF10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54253
Encompass_Preferred
Mouse Fibroblast Growth Factor 10 (FGF10) ELISA Kit for detecting FGF10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: O35565. Gene ID: 14165. Target Synonyms: Keratinocyte growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1239