Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 15 (FGF15) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54264
Encompass_Preferred
Mouse Fibroblast Growth Factor 15 (FGF15) ELISA Kit for detecting FGF15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: O35622. Gene ID: 14170. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1241