Learn More
Biomatik Corporation Human Fibroblast Growth Factor 9 (FGF9) ELISA Kit, 96 tests
Catalog No. 501501261
Encompass_Preferred
The Human Fibroblast Growth Factor 9 (FGF9) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.064 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P31371. NCBI Gene ID: 2254. Restriction: For Research Use Only.
Catalog No. 50-150-1261 Supplier Biomatik Corporation Supplier No. EKL54305
The Human Fibroblast Growth Factor 9 (FGF9) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.064 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P31371. NCBI Gene ID: 2254. Restriction: For Research Use Only.