Learn More
Biomatik Corporation Human Fibroblast Growth Factor Receptor 2 (FGFR2) ELISA Kit, 96 tests
Catalog No. 501501266
Encompass_Preferred
The Human Fibroblast Growth Factor Receptor 2 (FGFR2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.2 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P21802. NCBI Gene ID: 2263. Restriction: For Research Use Only.
Catalog No. 50-150-1266 Supplier Biomatik Corporation Supplier No. EKL54311
The Human Fibroblast Growth Factor Receptor 2 (FGFR2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.2 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P21802. NCBI Gene ID: 2263. Restriction: For Research Use Only.