Learn More
Biomatik Corporation Rat Fibronectin (FN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54317
Encompass_Preferred
Rat Fibronectin (FN) ELISA Kit for detecting FN in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.254ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P04937. Gene ID: 25661. Target Synonyms: FN1; CIG; FINC; LETS; MSF; GFND2; Anastellin; Migration-Stimulating Factor; Cold-Insoluble Globulin; Large; External; Transformation-Sensitive Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1271