Learn More
Biomatik Corporation Mouse Glucagon (GC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54328
Encompass_Preferred
Mouse Glucagon (GC) ELISA Kit for detecting GC in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 23.437-1500pg/ml. Sensitivity: 17.34pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P55095. Gene ID: 14526. Target Synonyms: GCG; GLP1; GLP2; GRPP; Glicentin-Related Polypeptide; Glucagen; Oxyntomodulin; Incretin hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1274