Learn More
Biomatik Corporation Rat Colony Stimulating Factor 3, Granulocyte (GCSF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54331
Encompass_Preferred
Rat Colony Stimulating Factor 3, Granulocyte (GCSF) ELISA Kit for detecting GCSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P97712. Gene ID: 25610. Target Synonyms: CSF3; G-CSF; Granulocyte Colony Stimulating Factor; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1277