Learn More
Biomatik Corporation Human Growth Differentiation Factor 10 (GDF10) ELISA Kit, 96 tests
Catalog No. 501501279
Encompass_Preferred
The Human Growth Differentiation Factor 10 (GDF10) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.052 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P55107. NCBI Gene ID: 2662. Restriction: For Research Use Only.
Catalog No. 50-150-1279 Supplier Biomatik Corporation Supplier No. EKL54333
The Human Growth Differentiation Factor 10 (GDF10) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.052 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P55107. NCBI Gene ID: 2662. Restriction: For Research Use Only.