Learn More
Biomatik Corporation Human Ghrelin (GHRL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54340
Encompass_Preferred
The Human Ghrelin (GHRL) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.047 ng/mL. Format: 96-Well Plate. Assay type: competitive inhibition. UniProt ID: Q9UBU3. NCBI Gene ID: 51738. Restriction: For Research Use Only.
Catalog No. 50-150-1282