Learn More
Biomatik Corporation General Hyaluronic Acid (HA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54361
Encompass_Preferred
The General Hyaluronic Acid (HA) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: All species. Detection Range: 6.25-400 ng/mL. Sensitivity: 2.17 ng/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-1288