Learn More
Biomatik Corporation Human Hepatoma Derived Growth Factor (HDGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54366
Encompass_Preferred
The Human Hepatoma Derived Growth Factor (HDGF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P51858. NCBI Gene ID: 3068. Restriction: For Research Use Only.
Catalog No. 50-150-1292