Learn More
Biomatik Corporation Mouse Hepatoma Derived Growth Factor (HDGF) ELISA Kit, 96 tests
Catalog No. 501501293
Encompass_Preferred
Mouse Hepatoma Derived Growth Factor (HDGF) ELISA Kit for detecting HDGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P51859. Gene ID: 15191. Target Synonyms: HMG1L2; High-Mobility Group Protein 1-Like 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1293 Supplier Biomatik Corporation Supplier No. EKL54367
Mouse Hepatoma Derived Growth Factor (HDGF) ELISA Kit for detecting HDGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P51859. Gene ID: 15191. Target Synonyms: HMG1L2; High-Mobility Group Protein 1-Like 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.