Learn More
Biomatik Corporation Human Insulin Like Growth Factor 1 (IGF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54426
Encompass_Preferred
The Human Insulin Like Growth Factor 1 (IGF1) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.625-40 ng/mL. Sensitivity: 0.276 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P05019. NCBI Gene ID: 3479. Restriction: For Research Use Only.
Catalog No. 50-150-1322