Learn More
Biomatik Corporation Rat Insulin Like Growth Factor 1 (IGF1) ELISA Kit, 96 tests
Catalog No. 501501324
Encompass_Preferred
Rat Insulin Like Growth Factor 1 (IGF1) ELISA Kit for detecting IGF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P08025. Gene ID: 24482. Target Synonyms: IGFI; IGF1A; IBP1; MGF; Somatomedin C; Mechano Growth Gactor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1324 Supplier Biomatik Corporation Supplier No. EKL54429
Rat Insulin Like Growth Factor 1 (IGF1) ELISA Kit for detecting IGF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P08025. Gene ID: 24482. Target Synonyms: IGFI; IGF1A; IBP1; MGF; Somatomedin C; Mechano Growth Gactor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.