Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor 2 (IGF2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54436
Encompass_Preferred
Mouse Insulin Like Growth Factor 2 (IGF2) ELISA Kit for detecting IGF2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 93.75-6000pg/ml. Sensitivity: 42.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P09535. Gene ID: 16002. Target Synonyms: INSIGF; IGF-II; Somatomedin A; Preptin; Insulin-like growth factor II Ala-25 Del. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1327