Learn More
Biomatik Corporation Rat Insulin Like Growth Factor 2 (IGF2) ELISA Kit, 96 tests
Catalog No. 501501328
Encompass_Preferred
Rat Insulin Like Growth Factor 2 (IGF2) ELISA Kit for detecting IGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.875-3000pg/ml. Sensitivity: 15.65pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P01346. Gene ID: 24483. Target Synonyms: INSIGF; IGF-II; Somatomedin A; Preptin; Insulin-like growth factor II Ala-25 Del. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1328 Supplier Biomatik Corporation Supplier No. EKL54438
Rat Insulin Like Growth Factor 2 (IGF2) ELISA Kit for detecting IGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.875-3000pg/ml. Sensitivity: 15.65pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P01346. Gene ID: 24483. Target Synonyms: INSIGF; IGF-II; Somatomedin A; Preptin; Insulin-like growth factor II Ala-25 Del. Restrictions: For Research Use Only. Not for use in diagnostic procedures.