Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 1 (IGFBP1) ELISA Kit, 96 tests
Catalog No. 501501332
Encompass_Preferred
Rat Insulin Like Growth Factor Binding Protein 1 (IGFBP1) ELISA Kit for detecting IGFBP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P21743. Gene ID: 25685. Target Synonyms: IBP1; AFBP; IGF-BP25; PP12; Placental Protein 12; Amniotic Fluid Binding Protein; Alpha-Pregnancy-Associated Endometrial Globulin; Growth Hormone Independent-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1332 Supplier Biomatik Corporation Supplier No. EKL54448
Rat Insulin Like Growth Factor Binding Protein 1 (IGFBP1) ELISA Kit for detecting IGFBP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P21743. Gene ID: 25685. Target Synonyms: IBP1; AFBP; IGF-BP25; PP12; Placental Protein 12; Amniotic Fluid Binding Protein; Alpha-Pregnancy-Associated Endometrial Globulin; Growth Hormone Independent-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.