Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54462
Encompass_Preferred
Mouse Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit for detecting IGFBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P47879. Gene ID: 16010. Target Synonyms: BP4; HT29-IGFBP; IBP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1341