Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54464
Encompass_Preferred
Rat Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit for detecting IGFBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.148ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P21744. Gene ID: 360622. Target Synonyms: BP4; HT29-IGFBP; IBP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1342