Learn More
Biomatik Corporation Human Interleukin 10 (IL10) ELISA Kit, 96 tests
Catalog No. 501501351
Encompass_Preferred
The Human Interleukin 10 (IL10) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 11.3 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P22301. NCBI Gene ID: 3586. Restriction: For Research Use Only.
Catalog No. 50-150-1351 Supplier Biomatik Corporation Supplier No. EKL54483
The Human Interleukin 10 (IL10) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 11.3 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P22301. NCBI Gene ID: 3586. Restriction: For Research Use Only.